Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCCTTACTTACTCAGTTCCTGC[C/T]GTAGCGTCTGGATGAGGGGGTATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600031 | ||||||||||||||||||||
Literature Links: |
CHIT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHIT1 - chitinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256125.1 | 1184 | Missense Mutation | CAG,CGG | Q,R 363 | NP_001243054.2 | |
NM_001270509.1 | 1184 | Missense Mutation | CAG,CGG | Q,R 337 | NP_001257438.1 | |
NM_003465.2 | 1184 | Missense Mutation | CAG,CGG | Q,R 382 | NP_003456.1 |