Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCCTGCGAGGTTTGGGACGGC[A/C]CCCGGCGCATGCCCGTTGAGCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616593 | ||||||||||||||||||||
Literature Links: |
FAM132A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM132A - family with sequence similarity 132 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2J2 - ubiquitin conjugating enzyme E2 J2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_058167.2 | 695 | Silent Mutation | GGG,GGT | G,G 211 | NP_477515.2 | |
NM_194315.1 | 695 | Silent Mutation | GGG,GGT | G,G 227 | NP_919296.1 | |
NM_194457.1 | 695 | Silent Mutation | GGG,GGT | G,G 159 | NP_919439.1 | |
NM_194458.1 | 695 | Silent Mutation | GGG,GGT | G,G 159 | NP_919440.1 | |
XM_005244718.3 | 695 | Silent Mutation | GGG,GGT | G,G 211 | XP_005244775.1 | |
XM_005244719.4 | 695 | Silent Mutation | GGG,GGT | G,G 211 | XP_005244776.1 | |
XM_006710333.3 | 695 | Silent Mutation | GGG,GGT | G,G 211 | XP_006710396.1 | |
XM_011540613.2 | 695 | Silent Mutation | GGG,GGT | G,G 176 | XP_011538915.1 | |
XM_011540614.2 | 695 | Silent Mutation | GGG,GGT | G,G 211 | XP_011538916.1 | |
XM_011540616.2 | 695 | Silent Mutation | GGG,GGT | G,G 159 | XP_011538918.1 | |
XM_011540617.2 | 695 | Silent Mutation | GGG,GGT | G,G 159 | XP_011538919.1 | |
XM_017000238.1 | 695 | Silent Mutation | GGG,GGT | G,G 159 | XP_016855727.1 | |
XM_017000239.1 | 695 | Silent Mutation | GGG,GGT | G,G 159 | XP_016855728.1 |