Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610162 MIM: 608676 | ||||||||||||||||||||
Literature Links: |
CCDC28B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC28B - coiled-coil domain containing 28B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301011.1 | 2246 | Intron | NP_001287940.1 | |||
NM_024296.4 | 2246 | Intron | NP_077272.2 | |||
XM_011542113.2 | 2246 | Intron | XP_011540415.1 | |||
XM_017002306.1 | 2246 | Intron | XP_016857795.1 | |||
XM_017002307.1 | 2246 | Intron | XP_016857796.1 | |||
XM_017002308.1 | 2246 | Intron | XP_016857797.1 | |||
XM_017002309.1 | 2246 | Intron | XP_016857798.1 |
DCDC2B - doublecortin domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IQCC - IQ motif containing C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160042.1 | 2246 | Silent Mutation | GCA,GCT | A,A 12 | NP_001153514.1 | |
NM_018134.2 | 2246 | Silent Mutation | GCA,GCT | A,A 12 | NP_060604.2 |
TXLNA - taxilin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |