Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGTCACGATGTCAGCGTTGGC[C/G]GCCTGCACTGCGATGGCCAACGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601328 | ||||||||||||||||||||
Literature Links: |
ACAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAP3 - ArfGAP with coiled-coil, ankyrin repeat and PH domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030649.2 | 4884 | Silent Mutation | GCC,GCG | A,A 779 | NP_085152.2 | |
XM_005244715.2 | 4884 | Silent Mutation | GCC,GCG | A,A 746 | XP_005244772.1 | |
XM_005244717.3 | 4884 | Silent Mutation | GCC,GCG | A,A 438 | XP_005244774.1 | |
XM_011540606.2 | 4884 | Silent Mutation | GCC,GCG | A,A 789 | XP_011538908.1 | |
XM_011540607.1 | 4884 | Silent Mutation | GCC,GCG | A,A 789 | XP_011538909.1 | |
XM_011540608.2 | 4884 | Silent Mutation | GCC,GCG | A,A 757 | XP_011538910.1 | |
XM_011540609.2 | 4884 | Silent Mutation | GCC,GCG | A,A 756 | XP_011538911.1 | |
XM_017000233.1 | 4884 | Silent Mutation | GCC,GCG | A,A 590 | XP_016855722.1 |
MIR6726 - microRNA 6726 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCNN1D - sodium channel epithelial 1 delta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |