Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTGCAGATGACCCAGAGGTGC[A/G]GGTTCTACATAGCATCGGTCACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605951 MIM: 191164 MIM: 613683 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPM3 - dolichyl-phosphate mannosyltransferase subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFNA1 - ephrin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004428.2 | 560 | Missense Mutation | CAG,CGG | Q,R 174 | NP_004419.2 | |
NM_182685.1 | 560 | Missense Mutation | CAG,CGG | Q,R 152 | NP_872626.1 |
SLC50A1 - solute carrier family 50 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |