Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTCCCTAGCACAGCGAGAGGC[C/T]GAGTACGCCGAGGCCCGGAAGCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA21 - spermatogenesis associated 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SZRD1 - SUZ RNA binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114600.2 | 507 | Silent Mutation | GCC,GCT | A,A 93 | NP_001108072.1 | |
NM_001271869.1 | 507 | Silent Mutation | GCC,GCT | A,A 92 | NP_001258798.1 |