Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGCCTGGGCTGTGTGGATTCG[C/T]ACATGCTGAGCCAGGCTGTTGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613484 MIM: 604084 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPEN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPEN - spen family transcriptional repressor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB17 - zinc finger and BTB domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242884.1 | 2435 | Silent Mutation | GTA,GTG | V,V 654 | NP_001229813.1 | |
NM_001287603.1 | 2435 | Silent Mutation | GTA,GTG | V,V 743 | NP_001274532.1 | |
NM_001287604.1 | 2435 | Silent Mutation | GTA,GTG | V,V 660 | NP_001274533.1 | |
NM_001324137.1 | 2435 | Silent Mutation | GTA,GTG | V,V 660 | NP_001311066.1 | |
NM_001324138.1 | 2435 | Silent Mutation | GTA,GTG | V,V 673 | NP_001311067.1 | |
NM_003443.2 | 2435 | Silent Mutation | GTA,GTG | V,V 736 | NP_003434.2 | |
XM_005245986.1 | 2435 | Silent Mutation | GTA,GTG | V,V 680 | XP_005246043.1 | |
XM_005245987.1 | 2435 | Silent Mutation | GTA,GTG | V,V 667 | XP_005246044.1 | |
XM_011542085.2 | 2435 | Silent Mutation | GTA,GTG | V,V 667 | XP_011540387.1 | |
XM_011542086.1 | 2435 | Silent Mutation | GTA,GTG | V,V 661 | XP_011540388.1 | |
XM_011542087.2 | 2435 | Silent Mutation | GTA,GTG | V,V 661 | XP_011540389.1 | |
XM_011542088.1 | 2435 | Silent Mutation | GTA,GTG | V,V 654 | XP_011540390.1 | |
XM_017002242.1 | 2435 | Silent Mutation | GTA,GTG | V,V 661 | XP_016857731.1 |