Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCATGCCCGCCACGCTGAGGCT[A/G]CTGATGCTCGTGCCCGAGGGCGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600210 | ||||||||||||||||||||
Literature Links: |
RUNX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RUNX3 - runt related transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031680.2 | 1281 | Silent Mutation | AGC,AGT | S,S 304 | NP_001026850.1 | |
NM_001320672.1 | 1281 | Silent Mutation | AGC,AGT | S,S 304 | NP_001307601.1 | |
NM_004350.2 | 1281 | Silent Mutation | AGC,AGT | S,S 290 | NP_004341.1 | |
XM_005246024.4 | 1281 | Silent Mutation | AGC,AGT | S,S 304 | XP_005246081.1 | |
XM_011542351.1 | 1281 | Silent Mutation | AGC,AGT | S,S 251 | XP_011540653.1 | |
XM_017002670.1 | 1281 | Intron | XP_016858159.1 |