Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCGGCCTCAACTGCCTGGGTG[C/T]CTGGATCAAGTGCGGCAGTGTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609144 MIM: 610864 | ||||||||||||||||||||
Literature Links: |
FLVCR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLVCR1 - feline leukemia virus subgroup C cellular receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014053.3 | 737 | Missense Mutation | GCC,GTC | A,V 188 | NP_054772.1 | |
XM_011509446.2 | 737 | Missense Mutation | GCC,GTC | A,V 188 | XP_011507748.1 | |
XM_011509447.2 | 737 | Missense Mutation | GCC,GTC | A,V 188 | XP_011507749.1 | |
XM_011509448.1 | 737 | Missense Mutation | GCC,GTC | A,V 188 | XP_011507750.1 |
FLVCR1-AS1 - FLVCR1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |