Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCTGCACTGAGTGGACTGTCT[A/G]TGTCTTGAGAGGGAGCTGCATTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf112 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf112 - chromosome 1 open reading frame 112 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL18 - methyltransferase like 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320199.1 | 483 | Missense Mutation | ACA,ATA | T,I 75 | NP_001307128.1 | |
NM_001320201.1 | 483 | Missense Mutation | ACA,ATA | T,I 75 | NP_001307130.1 | |
NM_033418.3 | 483 | Missense Mutation | ACA,ATA | T,I 75 | NP_219486.1 | |
XM_006711627.3 | 483 | Missense Mutation | ACA,ATA | T,I 75 | XP_006711690.1 |