Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACCCATCGAGGCCCCGACTTTC[A/G]GATCAGGGCTGGTTTGCAGGTGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609794 MIM: 602943 | ||||||||||||||||||||
Literature Links: |
LINGO4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINGO4 - leucine rich repeat and Ig domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004432.3 | 1472 | Nonsense Mutation | CGA,TGA | R,* 416 | NP_001004432.1 |
RORC - RAR related orphan receptor C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |