Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCTGGTGTCTTGGATTCCAAT[C/T]GCCAGCTGTGTGACCTGAGGCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALPL - alkaline phosphatase, liver/bone/kidney | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000478.5 | Intron | NP_000469.3 | ||||
NM_001127501.3 | Intron | NP_001120973.2 | ||||
NM_001177520.2 | Intron | NP_001170991.1 | ||||
XM_005245818.1 | Intron | XP_005245875.1 | ||||
XM_006710546.2 | Intron | XP_006710609.1 | ||||
XM_017000903.1 | Intron | XP_016856392.1 |