Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCAGAGCCGGGAACTAGAGGAC[A/G]GCCCGGCAGACATGCAGAAAGTACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
POGK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POGK - pogo transposable element with KRAB domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314014.1 | 565 | UTR 5 | NP_001300943.1 | |||
NM_017542.4 | 565 | Missense Mutation | AGC,GGC | S,G 29 | NP_060012.3 | |
XM_005245370.3 | 565 | Missense Mutation | AGC,GGC | S,G 29 | XP_005245427.1 | |
XM_011509814.2 | 565 | Missense Mutation | AGC,GGC | S,G 29 | XP_011508116.1 | |
XM_011509815.1 | 565 | Missense Mutation | AGC,GGC | S,G 29 | XP_011508117.1 | |
XM_017001914.1 | 565 | Missense Mutation | AGC,GGC | S,G 29 | XP_016857403.1 | |
XM_017001915.1 | 565 | UTR 5 | XP_016857404.1 |