Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTGGAGGATTTAATTCAATCAA[C/T]TTAGCAAGGCCCAAAATGAAACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600682 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC16A1 - solute carrier family 16 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166496.1 | 3490 | UTR 3 | NP_001159968.1 | |||
NM_003051.3 | 3490 | UTR 3 | NP_003042.3 | |||
XM_011542026.2 | 3490 | Intron | XP_011540328.1 | |||
XM_011542027.2 | 3490 | Intron | XP_011540329.1 |