Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACCAGGGTTGTCCCGCTGGCTT[C/G]GGTGGTCAACACGAATAGAGAGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603615 | ||||||||||||||||||||
Literature Links: |
LRRC41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC41 - leucine rich repeat containing 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006369.4 | 1723 | Missense Mutation | CCA,CGA | P,R 560 | NP_006360.3 | |
XM_017000074.1 | 1723 | Missense Mutation | CCA,CGA | P,R 568 | XP_016855563.1 |
RAD54L - RAD54-like (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |