Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGACGCCTCCCCTGGGTCAAGCT[C/T]CCCATTTACATCTTGGTGCAGTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606578 MIM: 605867 | ||||||||||||||||||||
Literature Links: |
AQP10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AQP10 - aquaporin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080429.2 | 340 | Silent Mutation | CTC,CTT | L,L 100 | NP_536354.2 | |
XM_011510104.2 | 340 | Silent Mutation | CTC,CTT | L,L 101 | XP_011508406.1 |
ATP8B2 - ATPase phospholipid transporting 8B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |