Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCACATTTTCCTGGAGACTGC[A/G]TCCTTGCGGGAGATTTGGTACTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609696 | ||||||||||||||||||||
Literature Links: |
ARID4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARID4B - AT-rich interaction domain 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206794.1 | 3722 | Missense Mutation | ACG,ATG | T,M 1188 | NP_001193723.1 | |
NM_016374.5 | 3722 | Missense Mutation | ACG,ATG | T,M 1188 | NP_057458.4 | |
NM_031371.3 | 3722 | Missense Mutation | ACG,ATG | T,M 1102 | NP_112739.2 | |
XM_006711781.2 | 3722 | Missense Mutation | ACG,ATG | T,M 1152 | XP_006711844.1 | |
XM_011544212.2 | 3722 | Missense Mutation | ACG,ATG | T,M 1188 | XP_011542514.1 | |
XM_017001468.1 | 3722 | Missense Mutation | ACG,ATG | T,M 1190 | XP_016856957.1 | |
XM_017001469.1 | 3722 | Missense Mutation | ACG,ATG | T,M 1154 | XP_016856958.1 | |
XM_017001470.1 | 3722 | Missense Mutation | ACG,ATG | T,M 1104 | XP_016856959.1 | |
XM_017001471.1 | 3722 | Intron | XP_016856960.1 | |||
XM_017001472.1 | 3722 | Missense Mutation | ACG,ATG | T,M 895 | XP_016856961.1 |