Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACATCTTGGATATTTTAACTGC[C/T]CTCTGTCACATTGCATTCTAAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F13B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F13B - coagulation factor XIII B chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001994.2 | 1954 | Missense Mutation | GAG,GGG | E,G 639 | NP_001985.2 | |
XM_011509283.2 | 1954 | Missense Mutation | GAG,GGG | E,G 639 | XP_011507585.1 | |
XM_011509284.2 | 1954 | Missense Mutation | GAG,GGG | E,G 638 | XP_011507586.1 | |
XM_011509286.2 | 1954 | Missense Mutation | GAG,GGG | E,G 591 | XP_011507588.1 |