Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGAACTTGCTGGCTTCGATCCT[A/G]CACCTTCAATCTCATTCTGACTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609591 | ||||||||||||||||||||
Literature Links: |
KIAA0907 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0907 - KIAA0907 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014949.3 | 1574 | Missense Mutation | GCA,GTA | A,V 502 | NP_055764.2 |
RIT1 - Ras like without CAAX 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA4 - small Cajal body-specific RNA 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA80E - small nucleolar RNA, H/ACA box 80E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |