Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAAAGCAGGCCCTCTGGCTCAC[C/G]AAGACCAAGTTAATAGAAGGCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611843 | ||||||||||||||||||||
Literature Links: |
CYB5RL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYB5RL - cytochrome b5 reductase like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL37 - mitochondrial ribosomal protein L37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016491.3 | 449 | Silent Mutation | ACC,ACG | T,T 124 | NP_057575.2 | |
XM_005270920.3 | 449 | Silent Mutation | ACC,ACG | T,T 124 | XP_005270977.1 |