Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGATTTTAAATAGATTGACATA[A/C]GGTTTAGCAGAGTAATAGCTCTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAB21L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAB21L3 - mab-21 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152367.2 | Intron | NP_689580.2 | ||||
XM_011540623.2 | Intron | XP_011538925.1 | ||||
XM_011540625.2 | Intron | XP_011538927.1 | ||||
XM_011540626.2 | Intron | XP_011538928.1 | ||||
XM_017000260.1 | Intron | XP_016855749.1 |