Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCAGAAGTTTCAGGAATGTCAT[C/T]TGCACCATATTGATGGAATTTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL11A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL11A1 - collagen type XI alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190709.1 | 5403 | Silent Mutation | CAA,CAG | Q,Q 1656 | NP_001177638.1 | |
NM_001854.3 | 5403 | Silent Mutation | CAA,CAG | Q,Q 1695 | NP_001845.3 | |
NM_080629.2 | 5403 | Silent Mutation | CAA,CAG | Q,Q 1707 | NP_542196.2 | |
NM_080630.3 | 5403 | Silent Mutation | CAA,CAG | Q,Q 1579 | NP_542197.3 | |
XM_017000334.1 | 5403 | Silent Mutation | CAA,CAG | Q,Q 1746 | XP_016855823.1 | |
XM_017000335.1 | 5403 | Silent Mutation | CAA,CAG | Q,Q 1744 | XP_016855824.1 | |
XM_017000336.1 | 5403 | Intron | XP_016855825.1 | |||
XM_017000337.1 | 5403 | Silent Mutation | CAA,CAG | Q,Q 1212 | XP_016855826.1 |