Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCAAACCAACTCAATGTGAAAC[C/T]CCACTGTTTCTCCGTGAGGATCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC35D1 - solute carrier family 35 member D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015139.2 | 4659 | UTR 3 | NP_055954.1 | |||
XM_006710478.1 | 4659 | UTR 3 | XP_006710541.1 | |||
XM_011541070.1 | 4659 | UTR 3 | XP_011539372.1 |