Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCAGCTCTACGCCCAAATCTG[C/T]CGCATCGTCTGCCGCCATGCCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600241 MIM: 605875 | ||||||||||||||||||||
Literature Links: |
GPR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR3 - G protein-coupled receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005281.3 | 750 | Silent Mutation | TGC,TGT | C,C 217 | NP_005272.1 |
WASF2 - WAS protein family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |