Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTCTTTGTCTTGTAGCTGCGGG[C/T]GCTTACCTGCCCCCCCTGCAGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606228 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGO1 - argonaute 1, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317122.1 | 221 | Silent Mutation | GGC,GGT | G,G 11 | NP_001304051.1 | |
NM_001317123.1 | 221 | UTR 5 | NP_001304052.1 | |||
NM_012199.4 | 221 | Silent Mutation | GGC,GGT | G,G 11 | NP_036331.1 | |
XM_011541236.2 | 221 | Silent Mutation | GGC,GGT | G,G 11 | XP_011539538.1 | |
XM_017001016.1 | 221 | Silent Mutation | GGC,GGT | G,G 11 | XP_016856505.1 | |
XM_017001017.1 | 221 | Silent Mutation | GGC,GGT | G,G 11 | XP_016856506.1 | |
XM_017001018.1 | 221 | Intron | XP_016856507.1 | |||
XM_017001019.1 | 221 | UTR 5 | XP_016856508.1 |