Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGAGGAGGAGGAGCGGGAGCAG[A/G]AGCCCCAGGAGGGGGGAGGCTGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INTS3 - integrator complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324475.1 | 14 | Missense Mutation | GAA,GGA | E,G 29 | NP_001311404.1 | |
NM_023015.4 | 14 | Missense Mutation | GAA,GGA | E,G 29 | NP_075391.3 | |
XM_005245459.2 | 14 | Missense Mutation | GAA,GGA | E,G 29 | XP_005245516.1 | |
XM_005245461.2 | 14 | Intron | XP_005245518.1 | |||
XM_006711490.3 | 14 | UTR 5 | XP_006711553.1 | |||
XM_011509906.2 | 14 | Intron | XP_011508208.1 | |||
XM_017002132.1 | 14 | Intron | XP_016857621.1 |