Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAAACAGCTAGTGTCTCCTGCAG[C/G]TGCGCTTCTTTCAAACACAACCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600563 | ||||||||||||||||||||
Literature Links: |
PTGFR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGFR - prostaglandin F receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000959.3 | 307 | Missense Mutation | GCT,GGT | A,G 14 | NP_000950.1 | |
NM_001039585.1 | 307 | Missense Mutation | GCT,GGT | A,G 14 | NP_001034674.1 | |
XM_017001873.1 | 307 | Missense Mutation | GCT,GGT | A,G 14 | XP_016857362.1 |