Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCGTTCCCTCTGCTGGAATCTTT[C/T]AAGAGAAAAAAAAAAGTATTAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM34 - RNA binding motif protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161533.1 | Intron | NP_001155005.1 | ||||
NM_015014.2 | Intron | NP_055829.2 | ||||
XM_011544133.1 | Intron | XP_011542435.1 | ||||
XM_017000721.1 | Intron | XP_016856210.1 |