Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGATTCTTCTGTAACATCAGA[C/T]AATGCCATGTCACGACGATAGCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612280 | ||||||||||||||||||||
Literature Links: |
FUCA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FUCA1 - fucosidase, alpha-L- 1, tissue | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000147.4 | 604 | Silent Mutation | TTA,TTG | L,L 312 | NP_000138.2 | |
XM_005245821.2 | 604 | Silent Mutation | TTA,TTG | L,L 187 | XP_005245878.1 | |
XM_011541167.2 | 604 | Silent Mutation | TTA,TTG | L,L 101 | XP_011539469.1 | |
XM_017000905.1 | 604 | Silent Mutation | TTA,TTG | L,L 211 | XP_016856394.1 |