Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGACACCTCAAAGCCAGGCACTC[C/T]GGCCAGGGCTGCTGCAATCTCCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608972 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRTC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRTC2 - CREB regulated transcription coactivator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181715.2 | 2068 | Missense Mutation | AGA,GGA | R,G 637 | NP_859066.1 | |
XM_005244946.1 | 2068 | Missense Mutation | AGA,GGA | R,G 642 | XP_005245003.1 | |
XM_005244949.2 | 2068 | Missense Mutation | AGA,GGA | R,G 529 | XP_005245006.1 | |
XM_017000575.1 | 2068 | Missense Mutation | AGA,GGA | R,G 564 | XP_016856064.1 | |
XM_017000576.1 | 2068 | Missense Mutation | AGA,GGA | R,G 559 | XP_016856065.1 | |
XM_017000577.1 | 2068 | Missense Mutation | AGA,GGA | R,G 541 | XP_016856066.1 | |
XM_017000578.1 | 2068 | Missense Mutation | AGA,GGA | R,G 536 | XP_016856067.1 | |
XM_017000579.1 | 2068 | Missense Mutation | AGA,GGA | R,G 431 | XP_016856068.1 |
DENND4B - DENN domain containing 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |