Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCCGAAGCACTGTGGCCCCTAC[A/G]TAGCATCGTGGAGTGGTCATACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614522 MIM: 603417 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL12 - kelch like family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303051.1 | Intron | NP_001289980.1 | ||||
NM_001303109.1 | Intron | NP_001290038.1 | ||||
NM_021633.3 | Intron | NP_067646.1 | ||||
XM_011509835.1 | Intron | XP_011508137.1 | ||||
XM_011509836.1 | Intron | XP_011508138.1 | ||||
XM_011509837.1 | Intron | XP_011508139.1 | ||||
XM_017001995.1 | Intron | XP_016857484.1 |
RABIF - RAB interacting factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |