Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACAAACCAACAAAAGCTTTATT[C/T]AAGAACAGGTGACATAGTTGAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600889 | ||||||||||||||||||||
Literature Links: |
CFHR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFHR2 - complement factor H related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312672.1 | 624 | Missense Mutation | TCA,TTA | S,L 107 | NP_001299601.1 | |
NM_005666.3 | 624 | Missense Mutation | TCA,TTA | S,L 231 | NP_005657.1 | |
XM_005245113.3 | 624 | Missense Mutation | TCA,TTA | S,L 166 | XP_005245170.1 | |
XM_011509458.2 | 624 | Missense Mutation | TCA,TTA | S,L 227 | XP_011507760.1 | |
XM_011509459.2 | 624 | Missense Mutation | TCA,TTA | S,L 211 | XP_011507761.1 | |
XM_011509460.2 | 624 | Missense Mutation | TCA,TTA | S,L 162 | XP_011507762.1 | |
XM_017001109.1 | 624 | Missense Mutation | TCA,TTA | S,L 172 | XP_016856598.1 |