Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTTCTTAGTAAAGTGGTAGA[A/G]TGTATAGAGCAAAGAGACTTTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611339 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG4C - autophagy related 4C cysteine peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032852.3 | Intron | NP_116241.2 | ||||
NM_178221.2 | Intron | NP_835739.1 | ||||
XM_005271288.2 | Intron | XP_005271345.1 | ||||
XM_011542313.2 | Intron | XP_011540615.1 | ||||
XM_017002600.1 | Intron | XP_016858089.1 | ||||
XM_017002601.1 | Intron | XP_016858090.1 |