Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACCCGGGAAGTCGGGGTCTCCT[C/T]CGCCTTCTGTCTTTCTGCGTCCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601924 MIM: 605254 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPA - coatomer protein complex subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098398.1 | 395 | UTR 5 | NP_001091868.1 | |||
NM_004371.3 | 395 | UTR 5 | NP_004362.2 |
NCSTN - nicastrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290184.1 | 395 | UTR 5 | NP_001277113.1 | |||
NM_001290186.1 | 395 | Silent Mutation | CTC,CTT | L,L 18 | NP_001277115.1 | |
NM_015331.2 | 395 | Silent Mutation | CTC,CTT | L,L 18 | NP_056146.1 | |
XM_005245053.4 | 395 | Silent Mutation | CTC,CTT | L,L 18 | XP_005245110.1 | |
XM_011509363.2 | 395 | Silent Mutation | CTC,CTT | L,L 18 | XP_011507665.1 |