Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGAGGGAGTTCTCAGTGCCCC[C/T]GGACAGGCCTCTCCAGCTTCACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601989 MIM: 607986 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
S100A13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
S100A13 - S100 calcium binding protein A13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S100A14 - S100 calcium binding protein A14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020672.2 | 509 | Missense Mutation | CAG,CGG | Q,R 102 | NP_065723.1 | |
XM_005245362.1 | 509 | Missense Mutation | CAG,CGG | Q,R 102 | XP_005245419.1 | |
XM_017001875.1 | 509 | Missense Mutation | CAG,CGG | Q,R 102 | XP_016857364.1 |
S100A16 - S100 calcium binding protein A16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |