Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAGATTGAGAACGTCATCTGG[A/G]TTGGTCCCAAAAATGCTCTTGCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600684 | ||||||||||||||||||||
Literature Links: |
LOC105371470 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105371470 - uncharacterized LOC105371470 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY9 - lymphocyte antigen 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033667.2 | 355 | Missense Mutation | ATT,GTT | I,V 81 | NP_001028839.1 | |
NM_001261456.1 | 355 | Missense Mutation | ATT,GTT | I,V 81 | NP_001248385.1 | |
NM_001261457.1 | 355 | Missense Mutation | ATT,GTT | I,V 81 | NP_001248386.1 | |
NM_002348.3 | 355 | Missense Mutation | ATT,GTT | I,V 81 | NP_002339.2 | |
XM_011509548.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_011507850.1 | |
XM_011509549.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_011507851.1 | |
XM_011509550.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_011507852.1 | |
XM_011509552.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_011507854.1 | |
XM_011509556.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_011507858.1 | |
XM_011509560.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_011507862.1 | |
XM_017001297.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_016856786.1 | |
XM_017001298.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_016856787.1 | |
XM_017001299.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_016856788.1 | |
XM_017001300.1 | 355 | Missense Mutation | ATT,GTT | I,V 95 | XP_016856789.1 | |
XM_017001301.1 | 355 | Missense Mutation | ATT,GTT | I,V 81 | XP_016856790.1 | |
XM_017001302.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_016856791.1 | |
XM_017001303.1 | 355 | Missense Mutation | ATT,GTT | I,V 128 | XP_016856792.1 | |
XM_017001304.1 | 355 | UTR 5 | XP_016856793.1 |