Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCAGCTCATCTTCTAGCCCAAGC[C/T]CTAGCTCCAATCCAGCTGCTGACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608972 | ||||||||||||||||||||
Literature Links: |
CRTC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRTC2 - CREB regulated transcription coactivator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181715.2 | 2111 | Missense Mutation | GAG,GGG | E,G 651 | NP_859066.1 | |
XM_005244946.1 | 2111 | Missense Mutation | GAG,GGG | E,G 656 | XP_005245003.1 | |
XM_005244949.2 | 2111 | Missense Mutation | GAG,GGG | E,G 543 | XP_005245006.1 | |
XM_017000575.1 | 2111 | Missense Mutation | GAG,GGG | E,G 578 | XP_016856064.1 | |
XM_017000576.1 | 2111 | Missense Mutation | GAG,GGG | E,G 573 | XP_016856065.1 | |
XM_017000577.1 | 2111 | Missense Mutation | GAG,GGG | E,G 555 | XP_016856066.1 | |
XM_017000578.1 | 2111 | Missense Mutation | GAG,GGG | E,G 550 | XP_016856067.1 | |
XM_017000579.1 | 2111 | Missense Mutation | GAG,GGG | E,G 445 | XP_016856068.1 |
DENND4B - DENN domain containing 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |