Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTACCTGGATTTAAGAACTCGGA[A/C]CACTTTGGACTGGTTTGTCCAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602458 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBPHL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYBPHL - myosin binding protein H like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SORT1 - sortilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205228.1 | 5476 | UTR 3 | NP_001192157.1 | |||
NM_002959.6 | 5476 | UTR 3 | NP_002950.3 | |||
XM_005271100.2 | 5476 | UTR 3 | XP_005271157.1 | |||
XM_005271101.2 | 5476 | UTR 3 | XP_005271158.1 | |||
XM_005271102.1 | 5476 | UTR 3 | XP_005271159.1 | |||
XM_006710812.1 | 5476 | UTR 3 | XP_006710875.1 |