Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTCGAAGCACGTCATTGTCCAG[C/G]TGCAGTTGGTCAAGTCCCTGCAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614264 MIM: 616041 MIM: 191523 | ||||||||||||||||||||
Literature Links: |
ARHGAP30 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP30 - Rho GTPase activating protein 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSTD1 - thiosulfate sulfurtransferase like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USF1 - upstream transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276373.1 | 1023 | Missense Mutation | CAC,CAG | H,Q 272 | NP_001263302.1 | |
NM_007122.4 | 1023 | Missense Mutation | CAC,CAG | H,Q 272 | NP_009053.1 | |
NM_207005.2 | 1023 | Missense Mutation | CAC,CAG | H,Q 213 | NP_996888.1 |