Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGCTGTGTATTTCCCATAGAA[C/T]CTTTCAGCTTTCTTGTTGAATTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYF2 - SYF2 pre-mRNA splicing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015484.4 | 601 | Silent Mutation | AGA,AGG | R,R 224 | NP_056299.1 | |
NM_207170.3 | 601 | Silent Mutation | AGA,AGG | R,R 182 | NP_997053.1 |