Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCTGCAAAGCTAGAGTTCCTCT[A/G]ATCTTAATTTAACATAGGCTGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCK - LCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042771.2 | Intron | NP_001036236.1 | ||||
NM_005356.4 | Intron | NP_005347.3 | ||||
XM_011541453.2 | Intron | XP_011539755.1 | ||||
XM_011541454.2 | Intron | XP_011539756.1 | ||||
XM_011541455.2 | Intron | XP_011539757.1 |