Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAGCCAGTGAGTGGTGCAGAG[A/C]TGGGCACTCTACGTCGAGGTGGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609473 | ||||||||||||||||||||
Literature Links: |
CGN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CGN - cingulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020770.2 | 238 | Missense Mutation | ATG,CTG | M,L 32 | NP_065821.1 | |
XM_005245365.4 | 238 | Missense Mutation | ATG,CTG | M,L 32 | XP_005245422.1 |