Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAGAGCATGGCCAGCCGGGAC[A/G]CCCGGTGCCCAGAATGGAAACCGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC27 - coiled-coil domain containing 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152492.2 | 316 | Missense Mutation | ACC,GCC | T,A 78 | NP_689705.2 | |
XM_011540780.1 | 316 | Missense Mutation | ACC,GCC | T,A 78 | XP_011539082.1 | |
XM_017000402.1 | 316 | Missense Mutation | ACC,GCC | T,A 78 | XP_016855891.1 | |
XM_017000403.1 | 316 | Missense Mutation | ACC,GCC | T,A 78 | XP_016855892.1 | |
XM_017000404.1 | 316 | Missense Mutation | ACC,GCC | T,A 78 | XP_016855893.1 | |
XM_017000405.1 | 316 | Missense Mutation | ACC,GCC | T,A 78 | XP_016855894.1 | |
XM_017000406.1 | 316 | UTR 5 | XP_016855895.1 |
TP73-AS1 - TP73 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |