Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCAGCCGCAGGCCTTGACCACC[A/G]TGTTGCGGTGCTTGCGCAGGATGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602284 MIM: 610289 MIM: 602435 | ||||||||||||||||||||
Literature Links: |
BMP8B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BMP8B - bone morphogenetic protein 8b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005271149.4 | 1350 | Intron | XP_005271206.1 | |||
XM_011542022.2 | 1350 | Intron | XP_011540324.1 | |||
XM_011542024.2 | 1350 | Missense Mutation | ACG,ATG | T,M 419 | XP_011540326.2 | |
XM_011542025.2 | 1350 | Intron | XP_011540327.1 | |||
XM_017002155.1 | 1350 | Intron | XP_016857644.1 | |||
XM_017002156.1 | 1350 | Intron | XP_016857645.1 |
OXCT2 - 3-oxoacid CoA-transferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIE - peptidylprolyl isomerase E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195007.1 | 1350 | Intron | NP_001181936.1 | |||
NM_001319293.1 | 1350 | Intron | NP_001306222.1 | |||
NM_006112.3 | 1350 | Intron | NP_006103.1 | |||
NM_203456.2 | 1350 | Intron | NP_982281.1 | |||
XM_006710289.2 | 1350 | Intron | XP_006710352.1 | |||
XM_006710290.3 | 1350 | Intron | XP_006710353.1 | |||
XM_011540501.1 | 1350 | Intron | XP_011538803.1 | |||
XM_017000051.1 | 1350 | Intron | XP_016855540.1 | |||
XM_017000052.1 | 1350 | Intron | XP_016855541.1 |