Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCTGACTTCCACTCTCCTAGC[C/T]GTCATTGTGGGTGTCATTTATTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNIH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNIH3 - cornichon family AMPA receptor auxiliary protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322302.1 | Intron | NP_001309231.1 | ||||
NM_001322303.1 | Intron | NP_001309232.1 | ||||
NM_001322304.1 | Intron | NP_001309233.1 | ||||
NM_001322305.1 | Intron | NP_001309234.1 | ||||
NM_152495.1 | Intron | NP_689708.1 |
WDR26 - WD repeat domain 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |