Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCCAGTTTTAGTGATTGTTCC[A/G]TAATGCAAACCTGATCATGTCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5F1 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf162 - chromosome 1 open reading frame 162 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300834.1 | Intron | NP_001287763.1 | ||||
NM_001300835.1 | Intron | NP_001287764.1 | ||||
NM_174896.3 | Intron | NP_777556.1 | ||||
XM_011540712.2 | Intron | XP_011539014.1 | ||||
XM_017000323.1 | Intron | XP_016855812.1 | ||||
XM_017000324.1 | Intron | XP_016855813.1 |