Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCCCTATCTGGCTGGGCGGAC[A/G]CTGGTAGGATTGCGGTGGAGCCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609183 | ||||||||||||||||||||
Literature Links: |
AURKAIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AURKAIP1 - aurora kinase A interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127229.1 | 481 | Missense Mutation | CGT,TGT | R,C 103 | NP_001120701.1 | |
NM_001127230.1 | 481 | Missense Mutation | CGT,TGT | R,C 103 | NP_001120702.1 | |
NM_017900.2 | 481 | Missense Mutation | CGT,TGT | R,C 103 | NP_060370.1 | |
XM_011541670.2 | 481 | Missense Mutation | CGT,TGT | R,C 132 | XP_011539972.1 |