Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTATTTTTCCATCAGTATGTGCT[A/G]AAGCTACAGATGTTGAAACACGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611503 | ||||||||||||||||||||
Literature Links: |
CENPL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPL - centromere protein L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127181.2 | 1541 | Missense Mutation | TCA,TTA | S,L 359 | NP_001120653.1 | |
NM_001171182.1 | 1541 | Missense Mutation | TCA,TTA | S,L 313 | NP_001164653.1 | |
NM_033319.3 | 1541 | Missense Mutation | TCA,TTA | S,L 313 | NP_201576.1 |