Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAGATATGTTCCAGGACATCTC[A/G]GACACCCAGCAGGGACAGTCAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVA1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EVA1B - eva-1 homolog B (C. elegans) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3D21 - SH3 domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162530.1 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | NP_001156002.1 | |
NM_024676.4 | 945 | Missense Mutation | CAG,CGG | Q,R 180 | NP_078952.4 | |
XM_011542153.2 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | XP_011540455.1 | |
XM_017002340.1 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | XP_016857829.1 | |
XM_017002341.1 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | XP_016857830.1 | |
XM_017002342.1 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | XP_016857831.1 | |
XM_017002343.1 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | XP_016857832.1 | |
XM_017002344.1 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | XP_016857833.1 | |
XM_017002345.1 | 945 | Missense Mutation | CAG,CGG | Q,R 291 | XP_016857834.1 | |
XM_017002346.1 | 945 | Intron | XP_016857835.1 |