Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCAGGGCTGCGTGGTAGTTTGT[A/G]TGGACAGCACGGCCCATGCCCTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606162 MIM: 612836 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PANK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PANK4 - pantothenate kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018216.2 | 2234 | Silent Mutation | CAC,CAT | H,H 739 | NP_060686.2 | |
XM_011541708.2 | 2234 | Intron | XP_011540010.2 | |||
XM_017001633.1 | 2234 | Silent Mutation | CAC,CAT | H,H 584 | XP_016857122.1 |
PLCH2 - phospholipase C eta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |